7-Isopropyl-1,4-dimethylazulene
Catalog # CSLO4252.1000
Supplier: CAESAR LORENTZ
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:198.31 g/mol
Merck Index:12,04581
Boiling Pt:309 °C (1013 hPa)
Melting Pt:133 °C
Density:0.9759 g/cm³ (18 °C)
EINECS:207-701-2
:GHS07
CAS Number:489-84-9
Formula:C₁₅H₁₈
MDL Number:MFCD00003811