4-Ethylmorpholine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-74-3
:
Formula:C₆H₁₃NO
MDL Number:MFCD00006177
:115.18 g/mol
Boiling Pt:139 °C (760 mmHg)
Melting Pt:–63 °C
Density:0.9 g/cm³
Flash Pt:27 °C
Storage Temperature:Ambient
EINECS:202-885-0
UN:2734
ADR:8,II
:GHS06
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4420 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:-46° to -50° (20°C, 589 nm) (c=1 in DMF)