Order Entry
Germany
ContactUsLinkComponent
4-Benzylpiperidine 98%
4-Benzylpiperidine 98%
Certificates
undefined
4-Benzylpiperidine 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5370 to 1.5390 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information