Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-2-pyrrolidone (NMP)
N-Methyl-2-pyrrolidone (NMP)
N-Methyl-2-pyrrolidone (NMP)
N-Methyl-2-pyrrolidone (NMP)
KRAE92-030.5000
:
KRAEMER MARTIN
:
N-Methyl-2-pyrrolidone (NMP)
KRAE92-030.5000
:
KRAEMER MARTIN
:
Pk:
5 L
Seq:
5190
:
:
99.13 g/mol
:
13,06140
:
202 °C (1013 hPa)
:
–24 °C
:
1.033 g/cm³ (20 °C)
:
91 °C
:
212-828-1
:
GHS08
:
GHS07
:
Ambient
:
872-50-4
:
C₅H₉NO
:
MFCD00003193