Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, 9,10-Diphenylanthracene
Supelco® Organic Reference Standard, 9,10-Diphenylanthracene
Catalog # 45788-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 9,10-Diphenylanthracene
Catalog # 45788-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1499-10-1
  • Description:
    9,10-Diphenylanthracene
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00001253
  • M-Clarity quality level:
    MQ100
  • Melting point:
    245 - 248 °C (lit.)
  • Molecular weight:
    330,42

Specifications

About this item