2-Butyne-1,4-diol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 110-65-6 Molecular Formula: C4H6O2 Formula Weight: 86.09 Flash Point: 152°(305°F) Melting Point: 54-58° Boiling Point: 194°/100mm Beilstein Reference: 1071237 UN No.: 2716 MDL No.: MFCD00002915
Synonyms: 1,4-Dihydroxy-2-butyne, But-2-yne-1,4-diol
- CAS Number:110-65-6
- Formula:C₄H₆O₂
- MDL Number:MFCD00002915
- Molecular Weight:86.09 g/mol
- Boiling Pt:194 °C (100 mmHg)
- Melting Pt:54…58 °C
- Density:1.1001 g/cm³ (20 °C)
- Flash Pt:152 °C (305 °F)
- Storage Temperature:Ambient
- EINECS:203-788-6
- UN:2716
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 110-65-6 Molecular Formula: C4H6O2 Formula Weight: 86.09 Flash Point: 152°(305°F) Melting Point: 54-58° Boiling Point: 194°/100mm Beilstein Reference: 1071237 UN No.: 2716 MDL No.: MFCD00002915
Synonyms: 1,4-Dihydroxy-2-butyne, But-2-yne-1,4-diol
- CAS Number:110-65-6
- Formula:C₄H₆O₂
- MDL Number:MFCD00002915
- Molecular Weight:86.09 g/mol
- Boiling Pt:194 °C (100 mmHg)
- Melting Pt:54…58 °C
- Density:1.1001 g/cm³ (20 °C)
- Flash Pt:152 °C (305 °F)
- Storage Temperature:Ambient
- EINECS:203-788-6
- UN:2716
- ADR:6.1,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



