3-Butyne-2-one
Catalog # MOLEM85823071
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:68.08 g/mol
Boiling Pt:85 °C (1013 hPa)
Density:0.886 g/cm³ (15 °C)
EINECS:215-834-2
UN:1992
ADR:3,II
CAS Number:1423-60-5
Formula:C₄H₄O
MDL Number:MFCD00008775