Order Entry
Germany
ContactUsLinkComponent
 
 
trans-4-Hydroxycinnamic acid ≥98% (by HPLC), Sigma-Aldrich®
 
undefined
 
trans-4-Hydroxycinnamic acid ≥98% (by HPLC), Sigma-Aldrich®

 

  • Appearance (Colour):
    White to Yellow and Faint Beige to Beige and Faint Brown to Brown
  • Appearance (Form):
    Powder
  • Purity (Hplc Area %):
    ≥98.0 %
  • Solubility (Colour):
    Colorless to Yellow
  • Solubility (Turbidity):
    Clear
  • Solubility (Method):
    50 mg/ml, Ethanol
  • Proton Nmr Spectrum:
    Conforms to structure