
trans-4-Hydroxycinnamic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:501-98-4
:7400-08-0
Formula:C₉H₈O₃
MDL Number:MFCD00004399
:164.16 g/mol
Merck Index:14,2560
Melting Pt:∼ 214 °C (dec.)
EINECS:605-511-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together