
trans-Cinnamic acid 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140-10-3
:621-82-9
Formula:C₉H₈O₂
MDL Number:MFCD00004369
Boiling Pt:300 °C (760 mmHg)
Melting Pt:131…136 °C
Density:1.248 g/cm³
Flash Pt:>100 °C
:148.16 g/mol
EINECS:205-398-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131 to 136 °C
- Titration with NaOH:≥98.0 %
- Elemental Analysis:45.67 to 48.01 % (C)