4-Chlorocinnamic acid
Glass bottle for solids, 100 g
Catalog # CHMPFL1144.100
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 3-(4-Chlorophenyl)acrylic acid
- CAS Number:1615-02-7
- Formula:C₉H₇ClO₂
- MDL Number:MFCD00004396
- Molecular Weight:182.61 g/mol
- Boiling Pt:311 °C (1013 hPa)
- Melting Pt:252…254 °C
- EINECS:216-564-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



