cis-9-Tricosene (muscalure), Sigma-Aldrich®
Catalog # SIAL859885-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:322.62 g/mol
Merck Index:12,06288
Boiling Pt:300 °C (1013 hPa)
Density:0.806 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:248-505-7
CAS Number:27519-02-4
Formula:C₂₃H₄₆
MDL Number:MFCD00008988