1-Dodecene 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-41-4
:
Formula:C₁₂H₂₄
MDL Number:MFCD00008961
:168.32 g/mol
Boiling Pt:212…213 °C
Melting Pt:–37 °C
Density:0.758
Flash Pt:73 °C (163 °F)
Storage Temperature:Ambient
EINECS:203-968-4
:GHS08
:GHS07