Methyl oleate
Catalog # EHERC15727060
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:296.49 g/mol
Merck Index:12,06965
Boiling Pt:∼218.5 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-992-5
CAS Number:112-62-9
:67762-38-3
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00009578