Order Entry
Germany
ContactUsLinkComponent
Bisacrylamide
Bisacrylamide
Catalog # UPTIUP864965
Supplier:  Uptima
CAS Number:  
undefined
Bisacrylamide
Catalog # UPTIUP864965
Supplier:  Uptima
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :154.17 g/mol
Boiling Pt:125 °C (3.33 hPa)
Melting Pt:185 °C
Density:1.235 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Refrigerator
EINECS:203-750-9
 :GHS07
CAS Number:110-26-9
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625