Order Entry
Germany
ContactUsLinkComponent
 
Bisacrylamide for synthesis, Sigma-Aldrich®
  8.05968.0250
 :  Merck
 :  
undefined
Bisacrylamide for synthesis, Sigma-Aldrich®
  8.05968.0250
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    185 °C (decomposition)
  • Molar Mass:
    154.17 g/mol
  • Bulk Density:
    200 kg/m3
  • Density:
    1.235 g/cm3 (30 °C)
  • pH:
    =5.0 (25 g/l, H2O, 20 °C)

 

 

 :
 :154.17 g/mol
 :125 °C (3.33 hPa)
 :185 °C
 :1.235 g/cm³ (20 °C)
 :138 °C
 :Refrigerator
 :203-750-9
 :2811
 :6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :110-26-9
 :(H₂C=CHCONH)₂CH₂
 :MFCD00008625