Bisacrylamide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-26-9
:
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625
:154.17 g/mol
Boiling Pt:125 °C (3.33 hPa)
Melting Pt:185 °C
Density:1.235 g/cm³ (20 °C)
Flash Pt:138 °C
UN:2811
ADR:6.1,
:M-Clarity™ quality level = MQ200
EINECS:203-750-9
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Assay (acidimetric following saponification) (%):min. 99,0
- DNases:not detectable
- Free acid (acrylic acid) (%):max. 0,02
- Heavy metals (as Pb) (%):max. 0,001
- Identity (IR-Spectrum):passes test
- Water (%):max. 0,5