Bisacrylamide, Sigma-Aldrich®
Catalog # 1.01546.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay (acidimetric following saponification) (%):min. 99,0
- DNases:not detectable
- Free acid (acrylic acid) (%):max. 0,02
- Identity (IR-Spectrum):passes test
- Proteases:not detectable
- RNases:not detectable
- Water (%):max. 0,5
Specification Test Results
About this item
:
:154.17 g/mol
Boiling Pt:125 °C (3.33 hPa)
Melting Pt:185 °C
Density:1.235 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Refrigerator
EINECS:203-750-9
UN:2811
ADR:6.1,II
:GHS06
:GHS08
CAS Number:110-26-9
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625