Bisacrylamide 99+% for biochemistry, for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-26-9
:
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625
:154.17 g/mol
Boiling Pt:125 °C (3,33 hPa)
Melting Pt:185 °C (dec.)
Density:1,235 g/cm³ (20 °C)
Flash Pt:138 °C
EINECS:203-750-9
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- HPLC:≥99.0 %
- UV:(1 % in water) 1 cm cell
- pH:5 to 7 (1 % in water)
- Acrylic acid:≤0.05 %