Order Entry
Germany
Orders LinkContactUsLinkComponent
Acrylamide, Millipore®

Acrylamide, Millipore®

Toxic
Health
Supplier:   Merck
Acrylamide, Millipore®
Supplier:  Merck
Certificates
Acrylamide, Millipore®
Acrylamide, Millipore®

2 Options Available Below

Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84.5 °C
  • Density:1.122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • ADR:6.1,
  •  :M-Clarity™ quality level = MQ200
  • UN:2074

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 53,60 €
    53,60 €
    1 * 100 g
       53,60 €

  • Glass bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 307,00 €
    307,00 €
    1 * 1 kg
       307,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.