Order Entry
Germany
ContactUsLinkComponent
 
HEPES sodium salt 99+% for molecular biology DNase-, RNase-, protease-free
 
undefined
HEPES sodium salt 99+% for molecular biology DNase-, RNase-, protease-free

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤3 % (K.F.)
  • UV:
    (0.1 M in water)
  • pH:
    9.5 to 11.5 (1 % in water)
  • Titration with HClO4:
    ≥99.0 % (on dry substance)
  • Dnase,Rnase,Protease activity:
    None detected