(4-Hydroxyphenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:156-38-7
:
Formula:(C₆H₄(OH))CH₂COOH
MDL Number:MFCD00004347
:152.15 g/mol
Boiling Pt:347 °C (1013 hPa)
Melting Pt:147…150 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-851-3
:GHS07