Order Entry
Germany
ContactUsLinkComponent
(4-Hydroxyphenyl)acetic acid
(4-Hydroxyphenyl)acetic acid
Catalog # MOLEM22647659
Supplier:  Molekula
CAS Number:  
(4-Hydroxyphenyl)acetic acid
Catalog # MOLEM22647659
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.15 g/mol
Boiling Pt:347 °C (1013 hPa)
Melting Pt:147…150 °C
EINECS:205-851-3
 :GHS07
Storage Temperature:Ambient
CAS Number:156-38-7
Formula:(C₆H₄(OH))CH₂COOH
MDL Number:MFCD00004347