Order Entry
Germany
ContactUsLinkComponent
 
 
3-Phenylpropionic acid 99%
 
undefined
 
3-Phenylpropionic acid 99%

 

  • Appearance:
    White to yellow to light beige Crystalline powder or crystals or mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    46 to 49 °C
  • GC:
    ≥98.5 %