Petrochemical standard, regular diesel fuel 2500 µg/ml in methanol, Dr. Ehrenstorfer
Catalog # EHERYA03004000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Concentration:2500 µg/ml in methanol
- Pk:1 mL
- Standards:Reference standard
- CAS No.:68334-30-5
- Description:Regular diesel fuel
Specifications
About this item
Certifications: ISO/IEC 17025 certified.