Order Entry
Germany
ContactUsLinkComponent
Crystal violet solution Grams for microscopy, Sigma-Aldrich®
Flammable
Irritant
Health
Crystal violet solution Grams for microscopy, Sigma-Aldrich®
Certificates
Flammable
Irritant
Health
Crystal violet solution Grams for microscopy, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (colour):
    Purple to very dark purple
  • Appearance (form):
    Liquid
  • Thin layer chromatogr.:
    Corresponds to standard
  • Density D20/4:
    0.970 - 0.985
  • Wavelength (1) (UV):
    585 - 595 nm
  • Extinction (1%/1 cm) (1) (UV):
    Report result
  • Solvent (UV):
    Water
  • Expiry period:
    36 months

Specification Test Results

Related Information

Frequently Bought Together