tert-Butyl methyl ether
Catalog # VWRS5562.9200
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:202 L
- Seq:5561
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
:GHS02
:GHS07
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812