L(-)-Glutathione (oxidised form)
Catalog # CAYM703216-1
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:612.64 g/mol
Melting Pt:185 °C
Storage Temperature:Refrigerator
EINECS:248-170-7
CAS Number:27025-41-8
Formula:C₂₀H₃₂N₆O₁₂S₂
MDL Number:MFCD00063106