Pivalamide, Sigma-Aldrich®
Catalog # SIALT71609-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:101.15 g/mol
Boiling Pt:212 °C (1013 hPa)
Density:1,02 g/cm³ (20 °C)
EINECS:212-043-4
CAS Number:754-10-9
Formula:C₅H₁₁NO
MDL Number:MFCD00008011