
Pivalamide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:754-10-9
:
Formula:C₅H₁₁NO
MDL Number:MFCD00008011
:101.15 g/mol
Boiling Pt:212 °C (760 mmHg)
Melting Pt:152…156 °C
Density:1,02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-043-4
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:152 to 156 °C
- GC:≥97.5 %