Pivaloyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3282-30-2
:
Formula:C₅H₉ClO
MDL Number:MFCD00000709
:120.58 g/mol
Boiling Pt:105 °C (1013 hPa)
Melting Pt:–56 °C
Density:0.979 g/cm³ (20 °C)
Flash Pt:8 °C
Storage Temperature:Ambient
UN:2438
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:221-921-6