n-Nonane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-84-2
:
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–53 °C
Density:0.718 g/cm³ (20 °C)
Flash Pt:31 °C
EINECS:203-913-4
UN:1920
ADR:3,III
:M-Clarity™ quality level = MQ100
:128.26 g/mol
:GHS02
:GHS08
Storage Temperature:Ambient