Order Entry
Germany
Orders LinkContactUsLinkComponent
n-Hexadecane, Sigma-Aldrich®

n-Hexadecane, Sigma-Aldrich®

Health
Supplier:   Merck
n-Hexadecane, Sigma-Aldrich®
Supplier:  Merck
Certificates
n-Hexadecane, Sigma-Aldrich®
n-Hexadecane, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Hexadecane, Cetane
  • CAS Number:544-76-3
  • Formula:H₃C(CH₂)₁₄CH₃
  • MDL Number:MFCD00008998
  • Molecular Weight:226.45 g/mol
  • Boiling Pt:287 °C (1013 hPa)
  • Melting Pt:18 °C
  • Density:0.77331 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:135 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:208-878-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 177,00 €
    177,00 €
    1 * 100 mL
       177,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 951,00 €
    951,00 €
    1 * 1 L
       951,00 €

  • Glass bottle
    2 L
    Log in for availability
    Log in for availability
    1 * 2 L
    1 * 2 L - 1 040,00 €
    1 040,00 €
    1 * 2 L
       1 040,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.