1-Butanesulphonic acid sodium salt 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2386-54-1
:
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540
:160.17 g/mol
Melting Pt:>300 °C
EINECS:219-201-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 % (1 g, 105°C, 2 h)
- Solubility:(10 % in water) Clear to slightly hazy colorless
- Titration after Ion exchange:≥97.5 %