2-Propanethiol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-33-2
:
Formula:C₃H₈S
MDL Number:MFCD00004863
:76.16 g/mol
Boiling Pt:57 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.81 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-861-4
ADR:3,II
:M-Clarity™ quality level = MQ300
UN:2402