ContactUsLinkComponent
1,6-Hexanediol, Sigma-Aldrich®
1,6-Hexanediol, Sigma-Aldrich®
  SIAL240117-50G
 :  Merck
 :  
1,6-Hexanediol, Sigma-Aldrich®
  SIAL240117-50G
 :  Merck
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368
 :
 :118.18 g/mol
 :13,04709
 :250 °C (1013 hPa)
 :39…42 °C
 :1.12 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :Ambient
 :211-074-0
 :629-11-8
 :C₆H₁₄O₂
 :MFCD00002985