1,3-Propanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:504-63-2
:
Formula:HO(CH₂)₃OH
MDL Number:MFCD00002949
:76.1 g/mol
Merck Index:12,09845
Boiling Pt:214 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.0597 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-997-3