Ethylene glycol analytical reagent
:107-21-1
:
:HOCH₂CH₂OH
:MFCD00002885
:62.07 g/mol
:13,03832
:196…198 °C (1013 hPa)
:–13 °C
:1.115 g/cm³ (20 °C)
:116 °C
:Refrigerator
:203-473-3
:01-2119456816-28
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion