
2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:59-52-9
:
Formula:C₃H₈OS₂
MDL Number:MFCD00004864
Merck Index:13,03234
Boiling Pt:223 °C (1013 hPa)
Melting Pt:77 °C
Density:1.2531 g/cm³ (20 °C)
Flash Pt:112 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:200-433-7
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:124.22 g/mol
:GHS06
Specification Test Results
- Solubility:87 g/l (20 °C) (slow decomposition)
- Molar Mass:124.22 g/mol
- Boiling Point:63 - 68 °C (0.27 hPa)
- Vapor Pressure:7.4 hPa (100 °C)
- Flash Point:112 °C
- Density:1.24 g/cm3 (20 °C)
- pH:5.0 - 6.5 (H2O, 20 °C) (saturated solution)
- Synonyms:Dithioglycerine, Dimercaprol, BAL
- Hill Formula:C3H8OS2
- Chemical Formula:HOCH2CH(SH)CH2SH