
2,3-Dimercapto-1-propanol 97.0-100.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59-52-9
:
Formula:C₃H₈OS₂
MDL Number:MFCD00004864
Merck Index:13,03234
Boiling Pt:120 °C (15 mmHg)
Melting Pt:77 °C
Density:1.243 g/cm³
Flash Pt:>112 °C
UN:2810
ADR:6.1,III
:124.22 g/mol
EINECS:200-433-7
:GHS06
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5700 to 1.5750 (20°C, 589 nm)
- Titration Iodimetric:97.0 to 100.5 %