

Ethanol (taxed) 96.0-97.2%, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
:64-17-5
:
:C₂H₅OH
:MFCD00003568
:46.07 g/mol
:78…79 °C (1.013 hPa)
:–115 °C
:0.805…0.812 g/cm3 (20 °C)
:12 °C
:Ambient
:200-578-6
:1170
:3,II
:GHS02
:GHS07
- Appearance:complying
- Identity A:complying
- Identity B:complying
- Boiling range:78 - 79 °C
- Density (D 20/20):0.805 - 0.812
- Water (Karl Fischer):4.8 - 7.4 %
- Non-volatile matter:Max. 0.001 %
- Acidity or alkalinity:complying
- Methanol (GC):Max. 200 Vol.ppm
- Volatile impurities (GC):complying
- Residual solvents:complying
- UV-absorption:complying
- Explosion Limit:19 %, 60 °F