2-Propanol
BSOL16264101BS
: BIOLAB
:
- Pack type:Glass bottle
- Pk:6 L
- Seq:5198
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
:
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS02
:GHS07
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674