1-Tetracosanol
Catalog # EHERC17396100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:190 °C (1013 hPa)
Melting Pt:81…83 °C
:354.66 g/mol
Storage Temperature:Ambient
EINECS:208-043-9
CAS Number:506-51-4
Formula:H₃C(CH₂)₂₂CH₂OH
MDL Number:MFCD00042662