1-Octanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-87-5
:
Formula:H₃C(CH₂)₆CH₂OH
MDL Number:MFCD00002988
:130.23 g/mol
Merck Index:13,06784
Boiling Pt:195 °C (1013 hPa)
Melting Pt:–16 °C
Density:0.8246 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:81 °C
EINECS:203-917-6
:GHS07
Storage Temperature:Ambient