1-Octacosanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:557-61-9
:
Formula:H₃C(CH₂)₂₆CH₂OH
MDL Number:MFCD00044770
:410.77 g/mol
Merck Index:12,06840
Boiling Pt:428 °C (1013 hPa)
Melting Pt:83.3 °C
:M-Clarity™ quality level = MQ200
EINECS:209-181-2