Order Entry
Germany
ContactUsLinkComponent
Acetic anhydride for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Toxic
Corrosive
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) Hydrolysis,Risk of violent reaction.
  • Melting Point:
    -73 °C
  • Molar Mass:
    102.08 g/mol
  • Boiling Point:
    138 - 140.5 °C (1013 hPa)
  • Vapor Pressure:
    4 hPa (20 °C)
  • Flash Point:
    49 °C
  • Explosion Limit:
    2.0 - 10.2 %(V)
  • Density:
    1.08 g/cm3 (20 °C)
  • pH:
    3 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    330 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together