Order Entry
Germany
ContactUsLinkComponent
 
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol), Millipore®
Catalog # 1.15118.9026
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol), Millipore®
Catalog # 1.15118.9026
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Receptacle PE/metal, returnable
  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :76.1 g/mol
Merck Index:12,06118
Boiling Pt:124.6 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.964 g/cm³ (20 °C)
Flash Pt:39 °C
Storage Temperature:Ambient
EINECS:203-713-7
UN:1188
ADR:3,
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-86-4
Formula:C₃H₈O₂
MDL Number:MFCD00002867