1,3-Diamino-2-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:616-29-5
:
Formula:C₃H₁₀N₂O
MDL Number:MFCD00008142
:90.13 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:40…42 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:210-474-2