Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Diamino-2-propanol
1,3-Diamino-2-propanol, Sigma-Aldrich®
1,3-Diamino-2-propanol, Sigma-Aldrich®
:
Merck
1,3-Diamino-2-propanol, Sigma-Aldrich®
:
Merck
:
616-29-5
:
:
C₃H₁₀N₂O
:
MFCD00008142
:
90.13 g/mol
:
237 °C (1013 hPa)
:
40…42 °C
:
M-Clarity™ quality level = MQ200
:
Refrigerator
:
210-474-2