Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane
- CAS Number:107-15-3
- Formula:C₂H₈N₂
- MDL Number:MFCD00008204
- Molecular Weight:60.1 g/mol
- Merck Index:13,03829
- Boiling Pt:117…118 °C (760 mmHg)
- Melting Pt:11 °C
- Density:0.898 g/cm³
- Flash Pt:38 °C
- Storage Temperature:Ambient
- EINECS:203-468-6
- UN:1604
- ADR:8,II
3 Options Available Below
Product Details & Documents
Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane
- CAS Number:107-15-3
- Formula:C₂H₈N₂
- MDL Number:MFCD00008204
- Molecular Weight:60.1 g/mol
- Merck Index:13,03829
- Boiling Pt:117…118 °C (760 mmHg)
- Melting Pt:11 °C
- Density:0.898 g/cm³
- Flash Pt:38 °C
- Storage Temperature:Ambient
- EINECS:203-468-6
- UN:1604
- ADR:8,II
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4555 to 1.4575 (20°C, 589 nm)
- Water:≤0.5 % (Coulometric)
- GC:≥98.5 %
- Sulfate (SO4):≤0.02 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤0.002 %
- Salicylic acid:≤0.04 % (HOC6H4COOH)
- Insoluble matter:≤0.02 %



